Få mere at vide
HG-9-91-01, MedChemExpress
Artikelnummer. 16459367
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16459367 5 mg -
16432897 50 mg 50mg
16416936 100 mg -
16416828 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16459367 Leverandør Medchem Express Leverandørnr. HY15776
MedChemExpress HG-9-91-01 is a potent and highly selective salt-inducible kinase (SIK) inhibitor with IC50s of 0.92 nM, 6.6 nM and 9.6 nM for SIK1, SIK2 and SIK3 respectively.
HG-9-91-01 is a potent and highly selective salt-inducible kinase (SIK) inhibitor with IC50s of 0.92 nM, 6.6 nM and 9.6 nM for SIK1, SIK2 and SIK3 respectively.
Tekniske data
Kemisk navn eller materiale HG-9-91-01
CAS 1456858-58-4
Farve White
Renhedsgrad noter Research
Molekylær formel C32H37N7O3
Mængde 5 mg
Synonym SIK inhibitor 1
Opløselighedsinformation DMSO : ≥ 150 mg/mL (264.23 mM)
SMIL CC1=CC=CC(C)=C1NC(N(C2=NC=NC(NC3=CC=C(N4CCN(C)CC4)C=C3)=C2)C5=CC=C(OC)C=C5OC)=O
Molekylvægt (g/mol) 567.68
Formel vægt 567.68
Procent renhed 98.02%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation