Få mere at vide
Hexadecamethylcyclooctasiloxane, TRC
Artikelnummer. 30134788
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134788 100 mg
30005344 50 mg
30004818 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134788 Leverandør Toronto Research Chemicals Leverandørnr. TRCH292510100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 556-68-3
Molekylær formel C16 H48 O8 Si8
Molekylvægt (g/mol) 593.23
Synonym 2,2,4,4,6,6,8,8,10,10,12,12,14,14,16,16-Hexadecamethylcyclooctasiloxane (ACI), Cyclooctasiloxane, hexadecamethyl- (7CI, 8CI, 9CI), Hexadecamethylcyclooctasiloxane, 2,2,4,4,6,6,8,8,10,10,12,12,14,14,16,16-Hexadecamethyl-1,3,5,7,9,11,13,15-octaoxa-2,4,6,8,10,12,14,16-octasilacyclohexadecane;
IUPAC navn 2,2,4,4,6,6,8,8,10,10,12,12,14,14,16,16-hexadecamethyl-1,3,5,7,9,11,13,15-octaoxa-2,4,6,8,10,12,14,16-octasilacyclohexadecane
SMIL C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1
Tekniske data
Kemisk navn eller materiale Hexadecamethylcyclooctasiloxane
Mængde 100 mg
InChI formel InChI=1S/C16H48O8Si8/c1-25(2)17-26(3,4)19-28(7,8)21-30(11,12)23-32(15,16)24-31(13,14)22-29(9,10)20-27(5,6)18-25/h1-16H3
Formel vægt 592.1503