missing translation for 'onlineSavingsMsg'
Learn More

Heptane, ≈99% N-Heptane, HPLC for Fluorescence Detection

800.00 DKK - 2185.00 DKK

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10684912
SDS View Documents
Fisher Chemical
H/0107/15
1 L
800.00 DKK
1 L
Please to purchase this item. Need a web account? Register with us today!
10355750
SDS View Documents
Fisher Chemical
H/0107/17
2.5 L
2185.00 DKK
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heptane, ≈99% N-Heptane, HPLC for Fluorescence Detection

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.