missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Hemoglobin A2 Antibody (rHBA/9167), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23944-100ug
This item is not returnable.
View return policy
Description
Hemoglobin A2 Monoclonal antibody specifically detects Hemoglobin A2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Hemoglobin A2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| alpha globin, alpha-2 globin, Alpha-globin, Hemoglobin alpha chain, hemoglobin subunit alpha, hemoglobin, alpha 2 | |
| Recombinant fragment (around aa1-100) of human Hemoglobin A2 protein (exact sequence is proprietary) | |
| 100 μg | |
| Cardiovascular Biology | |
| 3040 | |
| Store at 4°C. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| rHBA/9167 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction