Learn More
Miele™ Half Insert
Brand: Miele™ 3808710
Additional Details : Weight : 0.50000kg
Specifications
Insert, Half | |
18 Beakers up to 250mL | |
131 mm | |
1 Each |
E 144 | |
445 mm | |
200 mm |