Learn More
Miele™ Half Insert
1310.70 DKK - 3035.58 DKK
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
13152270
|
Miele™
3808330 |
Dimensions (D x W x H): 420 x 180 x 186mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13182270
|
Miele™
3808710 |
Dimensions (D x W x H): 445 x 200 x 131mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13162270
|
Miele™
3808390 |
Dimensions (D x W x H): 460 x 230 x 175mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Insert, Half | |
1 Each |