Få mere at vide
H-Ser-Tyr-OH, TRC
Artikelnummer. 30421422
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421422 50 mg
30417769 10 mg
30416254 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30421422 Leverandør Toronto Research Chemicals Leverandørnr. TRCS26205050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 21435-27-8
Molekylær formel C12H16N2O5
Molekylvægt (g/mol) 268.27
IUPAC navn (2S)-2-[[(2S)-2-amino-3-hydroxypropanoyl]amino]-3-(4-hydroxyphenyl)propanoic acid
SMIL N[C@@H](CO)C(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)O
Tekniske data
Kemisk navn eller materiale H-Ser-Tyr-OH
InChI formel InChI=1S/C12H16N2O5/c13-9(6-15)11(17)14-10(12(18)19)5-7-1-3-8(16)4-2-7/h1-4,9-10,15-16H,5-6,13H2,(H,14,17)(H,18,19)/t9-,10-/m0/s1
Mængde 50 mg
Formel vægt 268.1059