Få mere at vide
H-Gly-Pro-Ala-OH, TRC
Artikelnummer. 30240623
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240623 500 mg
30240710 100 mg
30240982 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30240623 Leverandør Toronto Research Chemicals Leverandørnr. TRCG650268500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 837-83-2
Molekylær formel C10H17N3O4
Molekylvægt (g/mol) 243.25
Synonym Glycylprolylalanine, Glycyl-L-prolyl-L-alanine, N-(1-Glycyl-L-prolyl)-L-alanine, N-(1-glycylprolyl)-L-alanine, ;
IUPAC navn (2S)-2-[[(2S)-1-(2-aminoacetyl)pyrrolidine-2-carbonyl]amino]propanoic acid
SMIL C[C@H](NC(=O)[C@@H]1CCCN1C(=O)CN)C(=O)O
Tekniske data
Kemisk navn eller materiale H-Gly-Pro-Ala-OH
InChI formel InChI=1S/C10H17N3O4/c1-6(10(16)17)12-9(15)7-3-2-4-13(7)8(14)5-11/h6-7H,2-5,11H2,1H3,(H,12,15)(H,16,17)/t6-,7-/m0/s1
Mængde 500 mg
Formel vægt 243.1219