Få mere at vide
GW284543, MedChemExpress
Artikelnummer. 16465596
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16465596 100 mg
16444798 10 mg
16411067 50 mg
16407547 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16465596 Leverandør Medchem Express Leverandørnr. HY114189
MedChemExpress GW284543 (UNC10225170) is a selective MEK5 inhibitor. GW284543 (UNC10225170) reduces pERK5, and decreases endogenous MYC protein.
GW284543 (UNC10225170) is a selective MEK5 inhibitor. GW284543 (UNC10225170) reduces pERK5, and decreases endogenous MYC protein.
Tekniske data
Kemisk navn eller materiale GW284543
CAS 790186-68-4
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C23H20N2O3
Mængde 100 mg
Synonym UNC10225170
Opløselighedsinformation DMSO : 125 mg/mL (335.64 mM; Need ultrasonic)
SMIL COC1=CC2=C(NC3=CC(OC4=CC=CC=C4)=CC=C3)C=CN=C2C=C1OC
Molekylvægt (g/mol) 372.42
Formel vægt 372.42
Procent renhed 99.92%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation