missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ GSTA4 (Human) Recombinant Protein

Human GSTA4 full-length ORF ( AAH15523, 1 a.a. - 222 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00002941-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135831

  • 4150.00 DKK / 25µg
Estimated Shipment: 16-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MAARPKLHYPNGRGRMESVRWVLAAAGVEFDEEFLETKEQLYKLQDGNHLLFQQVPMVEIDGMKLVQTRSILHYIADKHNLFGKNLKERTLIDMYVEGTLDLLELLIMHPFLKPDDQQKEVVNMAQKAIIRYFPVFEKILRGHGQSFLVGNQLSLADVILLQTILALEEKIPNILSAFPFLQEYTVKLSNIPTIKRFLEPGSKKKPPPDEIYVRTVYNIFRP
Specifications

Specifications

AAH15523
glutathione S-transferase alpha 4
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GSTA4
GST
2941
Wheat germ expression system
25 μg
DKFZp686D21185, GSTA4-4, GTA4
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers