Få mere at vide
GSK656, MedChemExpress
Artikelnummer. 16496857
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
Pakningsstørrelse:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16496857 5 mg -
16432976 25 mg -
16414158 10 mg -
16410507 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16496857 Leverandør Medchem Express Leverandørnr. HY107775
MedChemExpress GSK656 is a potent antitubercular agent, acting as an inhibitor of Mycobacterium tuberculosis (Mtb) leucyl-tRNA synthetase (LeuRS), with an IC50 of 0.2 μM.
GSK656 is a potent antitubercular agent, acting as an inhibitor of Mycobacterium tuberculosis (Mtb) leucyl-tRNA synthetase (LeuRS), with an IC50 of 0.2 μM.
Tekniske data
Kemisk navn eller materiale GSK656
CAS 2131798-13-3
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C10H14BCl2NO4
Mængde 5 mg
Opløselighedsinformation DMSO : 67.5 mg/mL (229.64 mM; Need ultrasonic)
SMIL OCCOC1=CC=C(Cl)C2=C1B(O)O[C@@H]2CN.[H]Cl
Molekylvægt (g/mol) 293.94
Formel vægt 293.94
Procent renhed 96.8%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation