Få mere at vide
Granzyme B (NK/T-Cell Lymphoma Marker) Mouse, Clone: GZMB/2403, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3002-MSM3-P0
Denne vare kan ikke returneres.
Se returpolitik
Tekniske data
| Granzyme B (NK/T-Cell Lymphoma Marker) | |
| Cytoplasmic | |
| GZMB/2403 | |
| Unconjugated | |
| Mouse | |
| 29-32kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 3002 | |
| IgG2b |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant fragment of human GZMB protein (apprx. aa 73-187) | |
| A431 cells. Tonsil, Spleen or Hodgkin's Lymphoma | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |