Learn More
Karl Hecht™ Erlenmeyer Flask, Standard Ground
Conical, flat-bottomed flask well suited to mixing by swirling. Karl Hecht™ Erlenmeyer Flask, Standard Ground is constructed of durable Duran™ borosilicate glass.
Brand:  Karl Hecht™ 41915012
 View more versions of this product
Product Code. 10585762
  • 158.00 DKK / Each
Only null left
Estimated Shipment: 06-08-2025
Description
  • Duran™ borosilicate glass.
  • Graduated.
  • Standard ground.
  • Reinforced ground neck marked with standard ground size.
  • Without stopper.
Specifications
Specifications
50 mL
Erlenmeyer
Yes
Borosilicate Glass (3.3), Duran Glass
NS 29/32
NS 29/32
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Karl Hecht™ Erlenmeyer Flask, Standard Ground > Capacity: 50mL