Learn More
Karl Hecht™ Erlenmeyer Flask, Standard Ground
Conical, flat-bottomed flask well suited to mixing by swirling. Karl Hecht™ Erlenmeyer Flask, Standard Ground is constructed of durable Duran™ borosilicate glass.
Brand: Karl Hecht™ 41915046
This item is not returnable.
View return policy
Description
- Duran™ borosilicate glass.
- Graduated.
- Standard ground.
- Reinforced ground neck marked with standard ground size.
- Without stopper.
Specifications
500 mL | |
Erlenmeyer | |
Yes |
Borosilicate Glass (3.3), Duran Glass | |
NS 45/40 | |
NS 45/40 |
Product Content Correction
Product Title
Karl Hecht™ Erlenmeyer Flask, Standard Ground >
Spot an opportunity for improvement?Share a Content Correction