Learn More
Karl Hecht™ Erlenmeyer Flask, Standard Ground
Conical, flat-bottomed flask well suited to mixing by swirling. Karl Hecht™ Erlenmeyer Flask, Standard Ground is constructed of durable Duran™ borosilicate glass.
Brand:  Karl Hecht™ 41915046
 View more versions of this product
Product Code. 10461132
  • 443.00 DKK / Each
Only null left
Estimated Shipment: 07-08-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Duran™ borosilicate glass.
  • Graduated.
  • Standard ground.
  • Reinforced ground neck marked with standard ground size.
  • Without stopper.
Specifications
Specifications
500 mL
Erlenmeyer
Yes
Borosilicate Glass (3.3), Duran Glass
NS 45/40
NS 45/40
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Karl Hecht™ Erlenmeyer Flask, Standard Ground >