Learn More
Lenz Laborglasinstrumente™ Graduated Cylindrical Separating Funnels with PTFE Plug
Made of borosilicate glass. Lenz Laborglasinstrumente™ Graduated Cylindrical Separating Funnels with PTFE Plug are excellent for liquid-liquid extractions and separating the components of a mixture into two immiscible solvent phases of different densities.
Brand:  Lenz Laborglasinstrumente™ 04026149
 View more versions of this product
Product Code. 10424291
  • 1070.00 DKK / Pack of 2
Only null left
Estimated Shipment: 14-10-2025
Description
  • With NS socket and PE stopper
  • With PTFE plug
  • With screwthread retaining device
  • Borosilicate glass
Specifications
Specifications
250 mL
Yes
4
Borosilicate Glass
10
2
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Graduated Cylindrical Separating Funnels with PTFE Plug > Capacity (Metric): 250mL