Learn More
Sartorius Stedim Biotech™ Grade 918 Smooth Papers for Qualitative and Technical Analyses, Discs
Sartorius Stedim Biotech™ Grade 918 Smooth Papers for Qualitative and Technical Analyses, Discs are suitable for analytical and technical purposes and feature a medium fast to slow flow rate.
Brand:  Sartorius Stedim Biotech™ FT-3-607-040
 View more versions of this product
Product Code. 15972970
  • 149.22 DKK / Pack of 100
Only null left
Estimated Shipment: 25-08-2025
Description
  • Made of refined pulp and cotton linters with > 95% alpha-cellulose
  • Medium fast to slow filtering
  • Non-sterile, bulk-packed
TRUSTED_SUSTAINABILITY
Specifications
Specifications
85 g/m2
Cotton Linters, Refined Pulp
45 sec.
100/Pk.
Qualitative Filter Paper
Disc
6 to 10 ÎĽm
918
0.17 mm
40 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius Stedim Biotech™ Grade 918 Smooth Papers for Qualitative and Technical Analyses, Discs > Diameter: 40 mm