Learn More
Sartorius Stedim Biotech™ Grade 918 Smooth Papers for Qualitative and Technical Analyses, Discs
Sartorius Stedim Biotech™ Grade 918 Smooth Papers for Qualitative and Technical Analyses, Discs are suitable for analytical and technical purposes and feature a medium fast to slow flow rate.
Brand: Sartorius Stedim Biotech™ FT-3-607-204
This item is not returnable.
View return policy
Description
- Made of refined pulp and cotton linters with > 95% alpha-cellulose
- Medium fast to slow filtering
- Non-sterile, bulk-packed
Specifications
85 g/m2 | |
Cotton Linters, Refined Pulp | |
45 sec. | |
100/Pk. | |
Qualitative Filter Paper |
Disc | |
6 to 10 ÎĽm | |
918 | |
0.17 mm | |
204 mm |
Product Content Correction
Product Title
Sartorius Stedim Biotech™ Grade 918 Smooth Papers for Qualitative and Technical Analyses, Discs > Diameter: 204 mm
Spot an opportunity for improvement?Share a Content Correction