Få mere at vide
Golgicide A, MedChemExpress
Artikelnummer. 16475807
Encompass_Preferred
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
16475807 5 mg
1 options
Artikelnummer. 16475807 Leverandør Medchem Express Leverandørnr. HY100540
MedChemExpress Golgicide A (GCA) is a potent, highly specific, and reversible inhibitor of the cis-Golgi ADP-ribosylation factor guanine nucleotide exchange factors (ArfGEF) GBF1. Golgicide A drastically reduced replication of coxsackievirus B3 (CVB3) and other human enterovirus species.
Golgicide A (GCA) is a potent, highly specific, and reversible inhibitor of the cis-Golgi ADP-ribosylation factor guanine nucleotide exchange factors (ArfGEF) GBF1. Golgicide A drastically reduced replication of coxsackievirus B3 (CVB3) and other human enterovirus species.
Tekniske data
Kemisk navn eller materiale Golgicide A
CAS 1139889-93-2
Farve White
Renhedsgrad noter Research
Molekylær formel C17H14F2N2
Mængde 5 mg
Synonym GCA
Opløselighedsinformation DMSO : 100 mg/mL (351.74 mM; Need ultrasonic)
SMIL FC1=CC2=C(NC(C3=CC=CN=C3)[C@@]4([H])[C@]2([H])C=CC4)C(F)=C1
Molekylvægt (g/mol) 284.3
Formel vægt 284.3
Procent renhed 98.79%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death