Få mere at vide
GlyH-101, MedChemExpress
Artikelnummer. 16453527
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16453527 50 mg
16449987 5 mg
16427508 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16453527 Leverandør Medchem Express Leverandørnr. HY18336
MedChemExpress GlyH-101 is a cell-permeable glycinyl hydrazone compound that blocks CFTR with Ki of 1.4 uM.
GlyH-101 is a cell-permeable glycinyl hydrazone compound that blocks CFTR with Ki of 1.4 uM.
Tekniske data
Kemisk navn eller materiale GlyH-101
CAS 328541-79-3
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C19H15Br2N3O3
Mængde 50 mg
Opløselighedsinformation DMSO : ≥ 58 mg/mL (117.61 mM)
SMIL O=C(N/N=C/C1=CC(Br)=C(O)C(Br)=C1O)CNC2=CC3=CC=CC=C3C=C2
Molekylvægt (g/mol) 493.15
Formel vægt 493.15
Procent renhed 98.24%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation