Få mere at vide
Glycyl-L-Glutamine, TRC
Artikelnummer. 30240776
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240776 1 mg
30240561 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30240776 Leverandør Toronto Research Chemicals Leverandørnr. TRCG5981251MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 211446-46-7
Molekylær formel C7 H13 N3 O4 . H2 O
Molekylvægt (g/mol) 221.21
Synonym Glycyl-L-glutamine hydrate
IUPAC navn (2S)-5-amino-2-[(2-aminoacetyl)amino]-5-oxopentanoic acid;hydrate
SMIL O.NCC(=O)N[C@@H](CCC(=O)N)C(=O)O
Tekniske data
Kemisk navn eller materiale Glycyl-L-Glutamine
InChI formel InChI=1S/C7H13N3O4.H2O/c8-3-6(12)10-4(7(13)14)1-2-5(9)11;/h4H,1-3,8H2,(H2,9,11)(H,10,12)(H,13,14);1H2/t4-;/m0./s1
Mængde 1 mg
Formel vægt 221.1012