Få mere at vide
Glycodeoxycholic Acid-3-O-Beta-glucuronide Disodium Salt, TRC
Artikelnummer. 30136569
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136569 5 mg
30134977 10 mg
30009257 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136569 Leverandør Toronto Research Chemicals Leverandørnr. TRCG6414055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 75672-36-5
Molekylær formel C32H49NNa2O11
Molekylvægt (g/mol) 669.71
Synonym Cholane, β-D-Glucopyranosiduronic Acid Derivative, (3α,5β,12α)-24-[(Carboxymethyl)amino]-12-hydroxy-24-oxocholan-3-yl, β-D-Glucopyranosiduronic Acid Disodium Salt
SMIL O=C(NCC(O[Na])=O)CC[C@@H](C)[C@](CC1)([H])[C@]2(C)[C@]1([H])[C@]3([H])CC[C@]4([H])C[C@H](O[C@H]5[C@H](O)C(O)[C@H](O)[C@@H](C(O[Na])=O)O5)CC[C@]4(C)[C@@]3([H])C[C@@H]2O
Tekniske data
Kemisk navn eller materiale Glycodeoxycholic Acid-3-O-beta-glucuronide Disodium Salt
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C32H51NO11.2Na/c1-15(4-9-23(35)33-14-24(36)37)19-7-8-20-18-6-5-16-12-17(10-11-31(16,2)21(18)13-22(34)32(19,20)3)43-30-27(40)25(38)26(39)28(44-30)29(41)42;;/h15-22,25-28,30,34,38-40H,4-14H2,1-3H3,(H,33,35)(H,36,37)(H,41,42);;/q;2*+1/p-2/t15-,16-,17-,18+,19-,20+,21+,22+,25?,26+,27-,28+,30-,31+,32-;;/m1../s1
Formel vægt 669.3101
Procent renhed >95%