missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerin, 100% Vegetable Based, Multi-Compendial, U.S.P., J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
500 mL
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail
Unit Size:
19 L
4 L
500mL
Specifications
Specifications
Flash Point | 199.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Quantity | 4 L |
Specific Gravity | 1L = 1.26kg |
Formula Weight | 92.1 |
Percent Purity | 100% Vegetable Based |
Grade | Multi-Compendial/USP |
Chemical Name or Material | Glycerin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Glycerin, 100% Vegetable Based, Multi-Compendial, U.S.P., J.T. Baker™ >
Spot an opportunity for improvement?Share a Content Correction