Få mere at vide
Gly-β-MCA, MedChemExpress
Artikelnummer. 16414828
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
Pakningsstørrelse:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16414828 10 mg -
16407577 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16414828 Leverandør Medchem Express Leverandørnr. HY114392
MedChemExpress Gly-β-MCA, a bile acid, is a potent, sable, intestine-selective and oral bioactive farnesoid X receptor (FXR) inhibitor that may be a candidate for the treatment of metabolic disorders.
Gly-β-MCA, a bile acid, is a potent, sable, intestine-selective and oral bioactive farnesoid X receptor (FXR) inhibitor that may be a candidate for the treatment of metabolic disorders.
Tekniske data
Kemisk navn eller materiale Gly-β-MCA
CAS 66225-78-3
Farve White
Renhedsgrad noter Research
Molekylær formel C26H43NO6
Mængde 10 mg
Opløselighedsinformation DMSO : 100 mg/mL (214.77 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMIL C[C@@H]([C@@]1([H])CC[C@@]2([H])[C@@]3([H])[C@]([C@]4(CC[C@@H](O)C[C@H]4[C@H](O)[C@@H]3O)C)([H])CC[C@]12C)CCC(NCC(O)=O)=O
Molekylvægt (g/mol) 465.62
Formel vægt 465.62
Procent renhed 98.0%
Grad Research
Fysisk form Powder
Til brug med (applikation) Metabolism-sugar/lipid metabolism