Learn More
DWK Life Sciences DURAN™ Adapter, stopper type
Compact reduction from sleeve to core.
Brand:  DWK Life Sciences 286576109
2246.75 DKK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 2.50000kg
 View more versions of this product
Product Code. 15864838
  • 2365.00 DKK / Pack of 10
Estimated Shipment: 10-06-2024
to see stock.
Add to basket
Description
Description
DURAN™ adapters are used to connect two components with different NS sizes. For the reduction from a larger core to a smaller core with low space requirements.
  • Compact reduction from sleeve to core.
  • Due to its compact design, this transition piece takes up very little space.
  • Stopper shape
Specifications
Specifications
10/Pk.
Connection of two components with different standard ground joint sizes
Solid
Socket NS 14/23, Cone NS 19/26