Learn More
DWK Life Sciences DURAN™ Adapter, stopper type
Compact reduction from sleeve to core.
180.76 DKK - 2650.00 DKK
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
15864838
|
DWK Life Sciences
286576109 |
Socket NS 14/23, Cone NS 19/26 |
2365.00 DKK
Pack of 10 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15874838
|
DWK Life Sciences
286576306 |
Socket NS 14/23, Cone NS 24/29 |
2465.00 DKK
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
15884838
|
DWK Life Sciences
286576709 |
Socket NS 19/26, Cone NS 24/29 |
2465.00 DKK
Pack of 10 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15894838
|
DWK Life Sciences
286576906 |
Socket NS 24/29, Cone NS 29/32 |
2650.00 DKK
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
15911621
|
DWK Life Sciences
286576803 |
Socket NS 19/26, Cone NS 29/32 |
2465.00 DKK
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
10131931
|
DWK Life Sciences
286576503 |
Socket NS 14/23, Cone NS 29/32 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
DURAN™ adapters are used to connect two components with different NS sizes. For the reduction from a larger core to a smaller core with low space requirements.
- Compact reduction from sleeve to core.
- Due to its compact design, this transition piece takes up very little space.
- Stopper shape
Specifications
10/Pk. | |
Solid |