Learn More
Karl Hecht™ Glass Centrifuge Tubes, Round Bottom
Clear, glass centrifuge tubes provide superior visibility. Karl Hecht™ Glass Centrifuge Tubes, Round Bottom have thick walls and are available graduated/ungraduated and constructed of AR™ soda glass or Duran™ borosilicate.
Brand:  Karl Hecht™ 40950004
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10481322
  • 22.31 DKK / Each
Estimated Shipment: 28-06-2024
to see stock.
Add to basket
Description
Description
  • Round bottom.
  • Graduated or ungraduated.
  • Rimless.
  • Thick-walled.
  • AR™ soda glass, or Duran™ borosilicate.
Specifications
Specifications
50 to 55 mL
Centrifuge
98 mm
33 to 35 mm
Centrifuge Tube
AR™ Soda Glass
50
Round