Learn More
Karl Hecht™ Glass Centrifuge Tubes, Round Bottom
Clear, glass centrifuge tubes provide superior visibility. Karl Hecht™ Glass Centrifuge Tubes, Round Bottom have thick walls and are available graduated/ungraduated and constructed of AR™ soda glass or Duran™ borosilicate.
Brand: Karl Hecht™ 40950002
Additional Details : Weight : 0.10000kg
Description
- Round bottom.
- Graduated or ungraduated.
- Rimless.
- Thick-walled.
- AR™ soda glass, or Duran™ borosilicate.
Specifications
10 to 15 mL | |
Centrifuge | |
107 mm | |
17 mm | |
Centrifuge Tube |
AR™ Soda Glass | |
100 | |
Round | |
No |