Få mere at vide
Gibberellin A6, TRC
Artikelnummer. 30240768
Skift visning
Mængde:
0.5 mg
1 mg
2.5 mg
Pakningsstørrelse:
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30240768 0.5 mg 0.5mg
30240752 2.5 mg 2.5mg
30240763 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product contains plant protection products according to Regulation (EC) No 1107/2009, all customers must complete a declaration before delivery.
Artikelnummer. 30240768 Leverandør Toronto Research Chemicals Leverandørnr. TRCG3773200.5MG
Regulatory: This product contains plant protection products according to Regulation (EC) No 1107/2009, all customers must complete a declaration before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 19147-78-5
Molekylær formel C19H22O6
Molekylvægt (g/mol) 346.37
Synonym GA6, 1,4a-Lactone-2beta,3beta-epoxy-4a,7-dihydroxy-1-methyl-8-methylene-4aalpha,4bbeta-gibbane-1alpha,10beta-dicarboxylic Acid;
SMIL C[C@]12[C@H]3O[C@H]3C[C@@]4(OC1=O)[C@@H]5CC[C@]6(O)C[C@]5(CC6=C)[C@H]([C@H]24)C(=O)O
Tekniske data
Kemisk navn eller materiale Gibberellin A6
Mængde 0.5 mg
InChI formel InChI=1S/C19H22O6/c1-8-5-17-7-18(8,23)4-3-10(17)19-6-9-13(24-9)16(2,15(22)25-19)12(19)11(17)14(20)21/h9-13,23H,1,3-7H2,2H3,(H,20,21)/t9-,10+,11+,12+,13-,16-,17-,18-,19+/m0/s1
Formel vægt 346.1416