Få mere at vide
IBA Lifesciences GFP-Strep-tag control protein Artikelnummer.: 15503077
IBA Lifesciences GFP-Strep-tag control protein
Artikelnummer. p-7230583 Shop alle IBA Lifesciences produkter
0.5 mL, 500µg
Mængde:
0.5 mL
5 mL
Pakningsstørrelse:
500µg
Artikelnummer. 15503077
Brand: IBA Lifesciences 21007005
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Se alternative produkter
GFP-Strep-tag control protein for Strep-Tactin™ and Strep-Tactin™XT applications
  • GFP control protein consisting of the green fluorescent protein (GFP) C-terminally tagged with the Strep-tag™II or Twin-Strep-tag™.
  • It is suitable for test purifications, detections, and immobilizations with Strep-Tactin™ or Strep-Tactin™XT.  
Tekniske data
Mængde 0.5 mL
Inkluderer 500 μg of a 30 kDa protein
Protein tag Twin-Strep-tag™
For Research Use Only.
Korrektion af produktindhold
Produkttitel
IBA Lifesciences GFP-Strep-tag control protein >