missing translation for 'onlineSavingsMsg'
Få mere at vide

GBR 12935 dihydrochloride, Thermo Scientific™

Artikelnummer. 15495269
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
Pakningsstørrelse:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15495269 50 mg 50mg
15485269 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15495269 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. J62991.MF
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

A very potent and selective inhibitor of dopamine uptake

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 67469-81-2
Molekylær formel C28H36Cl2N2O
Molekylvægt (g/mol) 487.509
MDL nummer MFCD00083175
InChI nøgle NQWRSILGEXNJIT-UHFFFAOYSA-N
Synonym gbr 12935 dihydrochloride, gbr-12935 dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-diphenylmethoxyethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-benzhydryloxyethyl-4-3-phenylpropyl piperazine;dihydrochloride, 1-2-benzhydryloxy ethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl-piperazine dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl piperazinediium dichloride, gbr12935 dihydrochloride
PubChem CID 11957553
ChEBI CHEBI:64091
IUPAC navn 1-(2-benzhydryloxyethyl)-4-(3-phenylpropyl)piperazine;dihydrochloride
SMIL C1CN(CCN1CCCC2=CC=CC=C2)CCOC(C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl

Tekniske data

Kemisk navn eller materiale GBR 12935 dihydrochloride
Farve White
Mængde 50 mg
Formel vægt 487.51
Fysisk form Powder
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.