Få mere at vide
Tocris Bioscience™ GBR 12783 dihydrochloride
GREENER_CHOICE
Artikelnummer. 18034472
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18034472 10 mg
18045352 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18034472 Leverandør Tocris Bioscience™ Leverandørnr. 0513/10
Potent and selective DA uptake inhibitor
A very potent and selective inhibitor of dopamine uptake (IC50 for inhibition of [3H]-dopamine uptake in rat striatal synaptosomes is 1.8 nM).
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 67469-75-4
Molekylær formel C28H34Cl2N2O
Tekniske data
Kemisk navn eller materiale GBR 12783 dihydrochloride
Mængde 10 mg
Formel vægt 485.5
Renhed >99%
Molekylvægt (g/mol) 485.493