Learn More
3M™ Gas, Vapour and Particulate Filter
3M™ Gas, Vapour and Particulate Filters have a bayonet-style connection, and they’re designed to optimise your field of vision. Our lightweight filters offer well-balanced breathing protection for hazardous environments.
36.15 DKK - 1210.00 DKK
Specifications
For Use With (Application) | Typical industrial applications for filters are laboratories and particulate applications |
---|---|
For Use With (Equipment) | 3M™ 6000 and 3M™ 7907 Series Full Face Masks |
Product Code | Brand | Model | Product Type | Filter Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Product Type | Filter Type | Price | Quantity & Availability | |||
17039263
|
3M™
7000021315 |
6098 | Gas, Vapor and Particulate Filter | AXP3 R |
835.00 DKK
2 pairs |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||
17113618
|
3M™
7000032083 |
6035 | Gas, Vapor and Particulate Filter | FFP3 |
1210.00 DKK
Pack of 20 |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||
17259996
|
3M™
7100066103 |
501 | Filter Retainer |
36.15 DKK
1 pair |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||||
15739007
|
3M™
7000030098 |
6096 | Gas, Vapor and Particulate Filter | A1 HgP3R |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Our lightweight filters combine protection with comfort. They offer low breathing resistance and good balance when connected to your mask, and they’re designed to optimise your field of vision. They can be clicked into place quickly and easily. Features a bayonet-style connection.Specifications
Typical industrial applications for filters are laboratories and particulate applications | |
3M™ 6000 and 3M™ 7907 Series Full Face Masks |