Få mere at vide
Tocris Bioscience™ GANT 61
GREENER_CHOICE
Artikelnummer. 18033992
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18033992 10 mg
18034332 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18033992 Leverandør Tocris Bioscience™ Leverandørnr. 3191/10
GLI antagonist; inhibits Hedgehog (Hh) signaling
GLI antagonist that inhibits GLI1 and GLI2-induced transcription (IC50 ∽ 5 μM). Inhibits the hedgehog (Hh) signaling pathway downstream of SMO and SUFU causing GLI1 nuclear accumulation. Displays antiproliferative and antitumor activity in vivo. Inhibits the pluripotency factors Oct4, Sox-2, Nanog and c-Myc in pancreatic cancer stem cells.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 500579-04-4
Molekylær formel C27H35N5
Tekniske data
Kemisk navn eller materiale GANT 61
Mængde 10 mg
Formel vægt 429.61
Renhed >98%
Molekylvægt (g/mol) 429.612