CAS RN 500579-04-4
IUPAC Navn:
2-[(3-{[2-(dimethylamino)phenyl]methyl}-2-(pyridin-4-yl)-1,3-diazinan-1-yl)methyl]-N,N-dimethylaniline
Synonymer:
2-[(3-{[2-(dimethylamino)phenyl]methyl}-2-(pyridin-4-yl)-1,3-diazinan-1-yl)methyl]-N,N-dimethylaniline
Molekylvægt (g/mol):
429.61
Molekylformel:
C27H35N5
InChi Key:
KVQOGDQTWWCZFX-UHFFFAOYSA-N
SMILES:
CN(C)C1=CC=CC=C1CN1CCCN(CC2=CC=CC=C2N(C)C)C1C1=CC=NC=C1
1
–
4
af
4
Resultater
1
Tocris Bioscience™ GANT 61
CAS: 500579-04-4 Molekylær formel: C27H35N5
2
GANT61, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4
GANT 61, MedChemExpress
MedChemExpress GANT 61 is an inhibitor of Gli1 and Gli2 targeting the Hedgehog/GLI pathway.