Få mere at vide
FOXE1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55363-1-AP-20UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The FOXE1 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human FOXE1. This antibody recognizes human, mouse antigen. The FOXE1 antibody has been validated for the following applications: IHC, ELISA analysis.Tekniske data
| FOXE1 | |
| Polyclonal | |
| Unconjugated | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Polyclonal Antibody | |
| IgG |
| Immunohistochemistry, ELISA | |
| 300 μg/mL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2304 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |