Få mere at vide
Formoterol O-Beta-D-Glucuronide (mixture of diastereomers), TRC
Artikelnummer. 30006235
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006235 1 mg
30009196 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30006235 Leverandør Toronto Research Chemicals Leverandørnr. TRCF6934101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 615551-59-2
Molekylær formel C25 H32 N2 O10
Molekylvægt (g/mol) 520.53
Synonym 2-(Formylamino)-4-[(1RS)-1-hydroxy-2-[[(1RS)-2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl β-D-Glucopyranosiduronic Acid
IUPAC navn (2S,3S,4S,5R,6S)-6-[2-formamido-4-[(1R)-1-hydroxy-2-[[(1R)-2-(4-methoxyphenyl)-1-methyl-ethyl]amino]ethyl]phenoxy]-3,4,5-trihydroxy-tetrahydropyran-2-carboxylic acid
SMIL COc1ccc(C[C@@H](C)NC[C@H](O)c2ccc(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)c(NC=O)c2)cc1
Tekniske data
Kemisk navn eller materiale Formoterol O-beta-D-Glucuronide (mixture of diastereomers)
Mængde 1 mg
InChI formel InChI=1S/C25H32N2O10/c1-13(9-14-3-6-16(35-2)7-4-14)26-11-18(29)15-5-8-19(17(10-15)27-12-28)36-25-22(32)20(30)21(31)23(37-25)24(33)34/h3-8,10,12-13,18,20-23,25-26,29-32H,9,11H2,1-2H3,(H,27,28)(H,33,34)/t13-,18+,20+,21+,22-,23+,25-/m1/s1
Formel vægt 520.206