Få mere at vide
Fmoc-t-butyl-L-alanine, TRC
Artikelnummer. 30009362
Skift visning
Mængde:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009362 250 mg
30008195 5 g
30007979 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009362 Leverandør Toronto Research Chemicals Leverandørnr. TRCF621070250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 139551-74-9
Molekylær formel C22 H25 N O4
Molekylvægt (g/mol) 367.44
Synonym Fmoc-neopentylglycine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4,4-dimethylpentanoic Acid, Fmoc-Ala(tBu)-OH, N-(9-Fluorenylmethyloxycarbonyl)-neopentylglycine
IUPAC navn (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4,4-dimethyl-pentanoic acid
SMIL CC(C)(C)C[C@H](NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O
Tekniske data
Kemisk navn eller materiale Fmoc-t-butyl-L-alanine
InChI formel InChI=1S/C22H25NO4/c1-22(2,3)12-19(20(24)25)23-21(26)27-13-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,18-19H,12-13H2,1-3H3,(H,23,26)(H,24,25)/t19-/m0/s1
Mængde 250 mg
Formel vægt 367.1784