Få mere at vide
Flumethasone, MedChemExpress
Artikelnummer. 16498576
Encompass_Preferred
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16498576 100 mg
16479875 250 mg
16414537 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16498576 Leverandør Medchem Express Leverandørnr. HYB1051
MedChemExpress Flumethasone is a corticosteroid for topical use, in combination with Clioquinol for the treatment of otitis externa and otomycosis.
Flumethasone is a corticosteroid for topical use, in combination with Clioquinol for the treatment of otitis externa and otomycosis.
Tekniske data
Kemisk navn eller materiale Flumethasone
CAS 2135-17-3
Farve White
Renhedsgrad noter Research
Molekylær formel C22H28F2O5
Mængde 100 mg
Synonym Flumetasone
Opløselighedsinformation DMSO : 100 mg/mL (243.64 mM; Need ultrasonic)
SMIL C[C@@]12[C@](C(CO)=O)(O)[C@H](C)C[C@@]1([H])[C@]3([H])C[C@H](F)C4=CC(C=C[C@]4(C)[C@@]3(F)[C@@H](O)C2)=O
Molekylvægt (g/mol) 410.45
Formel vægt 410.45
Procent renhed 99.63%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation