Learn More
Thermo Scientific™ Fluid Transfer Assembly, Extension Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Brand:  Thermo Scientific™ SH31117.01
 View more versions of this product
Product Code. 17810697
  • 10610.00 DKK / Pack of 10
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
  • Irradiation dose of 25-40 kGy provides minimum SAL of 10-6 for product contact surfaces
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6.35 mm (1/4 in.) ID; 11.11 mm (7/16 in.) OD; TPE 374; 1 m (3.25 ft); Luer
Extension Set
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Fluid Transfer Assembly, Extension Set > 6.35 mm (1/4 in.) ID; 11.11 mm (7/16 in.) OD; TPE 374; 1 m (3.25 ft); Luer