Learn More
Thermo Scientific™ Fluid Transfer Assembly, Adaptor Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Brand:  Thermo Scientific™ SH31122.01
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 17870647
  • 3705.00 DKK / Pack of 10
Estimated Shipment: 27-06-2024
to see stock.
Description
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Specifications
3.22 mm (1/8 in.) ID x 6.35 mm (1/4 in.) OD to 9.52 mm (3/8 in.) ID x 15.88 mm (5/8 in.) OD; TPE 374; 2 m (6.6 ft); Luer to MPC
Adapter Set