Få mere at vide
Fludrocortisone acetate, MedChemExpress
Beskrivelse
Fludrocortisone acetate (9α-Fludrocortisone acetate) is a synthetic mineralocorticoid, used to control the amount of sodium and fluids in your body.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Fludrocortisone acetate |
| CAS | 514-36-3 |
| Farve | White |
| Renhedsgrad noter | Research |
| Molekylær formel | C23H31FO6 |
| Mængde | 100 mg |
| Synonym | 9α-Fludrocortisone acetate 9α-Fluorcortisol acetate |
| Opløselighedsinformation | DMSO : 50 mg/mL (118.35 mM; Need ultrasonic) |
| SMIL | C[C@@]12[C@](C(COC(C)=O)=O)(O)CC[C@@]1([H])[C@]3([H])CCC4=CC(CC[C@]4(C)[C@@]3(F)[C@@H](O)C2)=O |
| Molekylvægt (g/mol) | 422.49 |
| Vis mere |