Få mere at vide
Flucarbazone-sodium, TRC
Artikelnummer. 30005232
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005232 250 mg
17950695 100 mg
17934855 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30005232 Leverandør Toronto Research Chemicals Leverandørnr. TRCF421125250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 181274-17-9
Molekylær formel C12 H10 F3 N4 O6 S . Na
Molekylvægt (g/mol) 418.28
Synonym Flucarbazone Sodium, 1H-1,2,4-Triazole-1-carboxamide, 4,5-dihydro-3-methoxy-4-methyl-5-oxo-N-[[2-(trifluoromethoxy)phenyl]sulfonyl]-, sodium salt (9CI), BAY-MKH 6562, Everest, Flucarbazone-sodium, MKH 6562, SJO 0498, 1H-1,2,4-Triazole-1-carboxamide, 4,5-dihydro-3-methoxy-4-methyl-5-oxo-N-[[2-(trifluoromethoxy)phenyl]sulfonyl]-, sodium salt (1:1)
IUPAC navn sodium;(3-methoxy-4-methyl-5-oxo-1,2,4-triazole-1-carbonyl)-[2-(trifluoromethoxy)phenyl]sulfonylazanide
SMIL [Na+].COC1=NN(C(=O)[N-]S(=O)(=O)c2ccccc2OC(F)(F)F)C(=O)N1C
Tekniske data
Kemisk navn eller materiale Flucarbazone-sodium
Mængde 250 mg
InChI formel InChI=1S/C12H11F3N4O6S.Na/c1-18-10(24-2)16-19(11(18)21)9(20)17-26(22,23)8-6-4-3-5-7(8)25-12(13,14)15;/h3-6H,1-2H3,(H,17,20);/q;+1/p-1
Formel vægt 418.0171