Få mere at vide
Flavanthrone, TRC
Artikelnummer. 30361852
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361852 50 mg
30352799 100 mg
30360051 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30361852 Leverandør Toronto Research Chemicals Leverandørnr. TRCC38225350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 475-71-8
Molekylær formel C28H12N2O2
Molekylvægt (g/mol) 408.41
Synonym Benzo[h]benz[5,6]acridino[2,1,9,8-klmna]acridine-8,16-dione
IUPAC navn 16,30-diazaoctacyclo[15.11.1.14,28.02,15.03,12.05,10.018,23.025,29]triaconta-1(28),2(15),3(12),4(30),5,7,9,13,16,18,20,22,25(29),26-tetradecaene-11,24-dione
SMIL O=C1c2ccccc2c3nc4ccc5C(=O)c6ccccc6c7nc8ccc1c3c8c4c57
Tekniske data
Kemisk navn eller materiale Flavanthrone
Mængde 50 mg
InChI formel InChI=1S/C28H12N2O2/c31-27-15-7-3-1-5-13(15)25-21-17(27)9-12-20-23(21)24-19(29-25)11-10-18-22(24)26(30-20)14-6-2-4-8-16(14)28(18)32/h1-12H
Formel vægt 408.0899