missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies FK-866

Nicotinamide phosphoribosyltransferase inhibitor

Encompass_Preferred

1540.35 DKK - 2624.40 DKK

Specifications

CAS 658084-64-1
Molecular Formula C24H29N3O2
Purity ≥98%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17179981
Encompass_Preferred
View Documents
STEMCELL Technologies
100-0263
5 mg
1540.35 DKK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17189981
Encompass_Preferred
View Documents
STEMCELL Technologies
100-0264
10 mg
2624.40 DKK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • FK-866 is a highly specific noncompetitive inhibitor of nicotinamide phosphoribosyltransferase (NAMPT), an enzyme that regulates NAD+ biosynthesis from the natural precursor nicotinamide (Cameron et al.; Hasmann & Schemainda)
  • In hepatocarcinoma cells, FK-866 activates AMP-activated protein kinase (AMPK) and downregulates mammalian target of rapamycin (mTOR) signaling (Schuster et al.)
  • FK-866 can be used to deplete NAD+, a central metabolism cofactor (Cameron et al.; Hasmann & Schemainda; Jadeja et al.)
  • Depletes NAD+ and induces delayed cell death by apoptosis in HepG2 human liver carcinoma cells (IC50 = ∼1 nM; Hasmann & Schemainda)
  • Triggers dose-dependent cytotoxicity in multiple myeloma cells (Cea et al.)
  • Induces autophagic death in neuroblastoma SH-SY5Y cells (IC50 = 0.93 nM; Billington et al.)
Specifications

Specifications

658084-64-1
C24H29N3O2
≥98%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
STEMCELL Technologies FK-866

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.