Learn More
Fisherbrand™ Color Coders for Externally and Internally Threaded Cryogenic Storage Vials
For sample identification
Brand: Fisherbrand™
559.92 DKK valid until 2024-04-30
Use promo code "21749" to get your promotional price.
Additional Details : Weight : 0.08400kg
Description
Fit into the caps of Fisherbrand Externally Threaded Cryogenic Storage Vials (10-500-25, -26, and -27) or Internally Threaded Cryogenic Storage Vials (12-567-500, -501, -502).
- For color-coded sample identification
- Polystyrene
- Nonsterile
Specifications
White | |
Polystyrene | |
Non-sterile | |
For externally threaded vials |