Learn More
Fisherbrand™ Color Coders for Externally and Internally Threaded Cryogenic Storage Vials
For sample identification
149.00 DKK - 695.00 DKK
Specifications
Material Polystyrene
Sterility Non-sterile
View More Specs
Products 10
Product Code Brand Color For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Color For Use With (Equipment) Price Quantity & Availability  
11927974
View Documents Promotion Details
Fisherbrand™
White For externally threaded vials
685.00 DKK
Pack of 500
View alternative products
Save Up to 
»
Estimated Shipment: 23-07-2024
to see stock.
Add to basket
11937974
View Documents Promotion Details
Fisherbrand™
Yellow For externally threaded vials
695.00 DKK
Pack of 500
View alternative products
Save Up to 
»
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
11947974
View Documents Promotion Details
Fisherbrand™
Green For externally threaded vials
690.00 DKK
Pack of 500
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11957974
View Documents Promotion Details
Fisherbrand™
Blue For externally threaded vials
695.00 DKK
Pack of 500
View alternative products
Save Up to 
»
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
11967974
View Documents Promotion Details
Fisherbrand™
Red For externally threaded vials
695.00 DKK
Pack of 500
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11977974
View Documents Promotion Details
Fisherbrand™
White For internally threaded vials
149.00 DKK
Pack of 500
View alternative products
Save Up to 
»
Estimated Shipment: 27-09-2024
to see stock.
Add to basket
11987974
View Documents Promotion Details
Fisherbrand™
Yellow For internally threaded vials
154.00 DKK
Pack of 500
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11997974
View Documents Promotion Details
Fisherbrand™
Green For internally threaded vials
164.00 DKK
Pack of 500
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11907984
View Documents Promotion Details
Fisherbrand™
Blue For internally threaded vials
149.00 DKK
Pack of 500
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11917984
View Documents Promotion Details
Fisherbrand™
Red For internally threaded vials
154.00 DKK
Pack of 500
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
Description
Description
Fit into the caps of Fisherbrand Externally Threaded Cryogenic Storage Vials (10-500-25, -26, and -27) or Internally Threaded Cryogenic Storage Vials (12-567-500, -501, -502).
  • For color-coded sample identification
  • Polystyrene
  • Nonsterile
Specifications
Specifications
Polystyrene