Learn More
Thermo Scientific™ Filters and Accessories for Thermo Scientific Fluoroskan™, Fluoroskan™ FL and Luminoskan™ Microplate Readers
Fluoroskan™, Fluoroskan™FL, and Luminoskan™ microplate fluorometers and luminometers can be equipped with a wide range of accessories, additional excitation filters, and emission filters to expand the instrument's functionality and application flexibility.
1854.00 DKK - 7900.00 DKK
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
15863537
|
Thermo Scientific™
11272700 |
1854.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12433398
|
Thermo Scientific™
1423202 |
320 nm |
4010.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12443398
|
Thermo Scientific™
1423352 |
335 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12463398
|
Thermo Scientific™
142340210 |
340 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12473398
|
Thermo Scientific™
142350220 |
350 nm |
7880.00 DKK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13343087
|
Thermo Scientific™
1423552 |
355 nm |
4000.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12493398
|
Thermo Scientific™
1423602 |
360 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12403408
|
Thermo Scientific™
142370212 |
370 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12413408
|
Thermo Scientific™
1423802 |
380 nm |
7860.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12433408
|
Thermo Scientific™
142395240 |
395 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10221300
|
Thermo Scientific™
1424002 |
400 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13363087
|
Thermo Scientific™
1424052 |
405 nm |
4015.00 DKK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12443408
|
Thermo Scientific™
142405210 |
405 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11859500
|
Thermo Scientific™
1424202 |
420 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12473408
|
Thermo Scientific™
1424252 |
425 nm |
7880.00 DKK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12483408
|
Thermo Scientific™
142430210 |
430 nm |
3985.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12719516
|
Thermo Scientific™
142440210 |
440 nm |
3985.00 DKK
Each |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12403418
|
Thermo Scientific™
1424442 |
444 nm |
3985.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12413418
|
Thermo Scientific™
1424502 |
450 nm |
7880.00 DKK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12423418
|
Thermo Scientific™
1424602 |
460 nm |
4005.00 DKK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17128595
|
Thermo Scientific™
142470L |
470 nm |
7090.00 DKK
Each |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12433418
|
Thermo Scientific™
142475210 |
475 nm |
7860.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12443418
|
Thermo Scientific™
1424802 |
480 nm |
7900.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11753037
|
Thermo Scientific™
1424852 |
485 nm |
4000.00 DKK
Each |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12463418
|
Thermo Scientific™
142485216 |
485 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12739516
|
Thermo Scientific™
1424902 |
490 nm |
7880.00 DKK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12483418
|
Thermo Scientific™
1424922 |
492 nm |
7880.00 DKK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11541559
|
Thermo Scientific™
1425102 |
510 nm |
4010.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11714958
|
Thermo Scientific™
1425152 |
515 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12493418
|
Thermo Scientific™
1425172 |
517 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11869500
|
Thermo Scientific™
1425182 |
518 nm |
4005.00 DKK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12403428
|
Thermo Scientific™
1425202 |
520 nm |
4000.00 DKK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12413428
|
Thermo Scientific™
1425212 |
521 nm |
7880.00 DKK
Each |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17138595
|
Thermo Scientific™
142525L |
525 nm |
7090.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11834383
|
Thermo Scientific™
1425272 |
527 nm |
3985.00 DKK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11712318
|
Thermo Scientific™
1425302 |
530 nm |
3915.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11919426
|
Thermo Scientific™
1425382 |
538 nm |
4005.00 DKK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12433428
|
Thermo Scientific™
142540210 |
540 nm |
7860.00 DKK
Each |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12443428
|
Thermo Scientific™
1425422 |
542 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12453428
|
Thermo Scientific™
1425442 |
544 nm |
3900.00 DKK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12463428
|
Thermo Scientific™
142550208 |
550 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11884750
|
Thermo Scientific™
1425552 |
555 nm |
3985.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11894303
|
Thermo Scientific™
142560210 |
560 nm |
7860.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12473428
|
Thermo Scientific™
142570210 |
570 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12483428
|
Thermo Scientific™
1425752 |
575 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11817993
|
Thermo Scientific™
142578206 |
578 nm |
4010.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12403438
|
Thermo Scientific™
1425842 |
584 nm |
4000.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12413438
|
Thermo Scientific™
1425902 |
590 nm |
3910.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12423438
|
Thermo Scientific™
1425972 |
597 nm |
7900.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12433438
|
Thermo Scientific™
1426032 |
603 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11827993
|
Thermo Scientific™
1426042 |
604 nm |
4000.00 DKK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12453438
|
Thermo Scientific™
1426122 |
612 nm |
4010.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11741238
|
Thermo Scientific™
1426202 |
620 nm |
4010.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11731238
|
Thermo Scientific™
142620220 |
620 nm |
7850.00 DKK
Each |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12463438
|
Thermo Scientific™
1426212 |
621 nm |
7880.00 DKK
Each |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12473438
|
Thermo Scientific™
142635255 |
635 nm |
7860.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12483438
|
Thermo Scientific™
1426402 |
640 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17148595
|
Thermo Scientific™
142640L |
640 nm |
7090.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12493438
|
Thermo Scientific™
1426462 |
646 nm |
3980.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12403448
|
Thermo Scientific™
142650220 |
650 nm |
7850.00 DKK
Each |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12413448
|
Thermo Scientific™
1426702 |
670 nm |
7860.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12423448
|
Thermo Scientific™
142678210 |
678 nm |
4010.00 DKK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12433448
|
Thermo Scientific™
1426802 |
680 nm |
3985.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12443448
|
Thermo Scientific™
1427002 |
700 nm |
7880.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12453448
|
Thermo Scientific™
1427902 |
790 nm |
7880.00 DKK
Each |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12453408
|
Thermo Scientific™
1424102 |
410 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12463408
|
Thermo Scientific™
142415210 |
415 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
For running fluorometric or luminometric assays, the Fluoroskan, Fluoroskan FL, and Luminoskan microplate readers offer reliability and optical technology that has already been proven in thousands of laboratories all over the world. Extensive choice of filters for optimal wavelength selection for assays increases the application flexibility. Additionally, the Thermo Scientific™ SkanIt™ Software increases efficiency in your lab.
- Filters play an essential role in fluorometric measurements
- When ordering special filters please provide as much information as possible for the best measurement results
- If advice is needed which filters to order, contact a local Thermo Fisher Scientific representative with the following details:
- Name of the application
- Excitation and emission spectra
- Peak or center wavelengths of the excitation and emission spectra
- Fluorometric filters can also be used for luminometry depending on the application
- For luminometric applications requiring filters, contact technical support for instructions