missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Filter

Use these condensers with Magnum Series compound microscopes. Fisherbrand™ filters are ergonomically designed for everyday use.

Brand:  Fisherbrand™

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11502663

  • 149.00 DKK / Each
Estimated Shipment: 30-07-2024
to see stock.
Add to basket

Description

Description

Magnum series microscopes have a robust T-shaped base, designed for maximum stability, with modern ergonomic features to ensure a comfortable, prolonged viewing position.
  • The trinocular versions allow connection to cameras whilst the planachromatic objectives provide true colour reproduction excluding spherical aberration
  • The phase contrast models allow an efficient view of thin or translucent specimens where staining is difficult
  • Capable of Kohler illumination, accessories are also available for darkfield and polarization enabling a wide range of applications
  • Head is inclined at 30° with interpupillary distance adjustment (Siedentopf type) of 55 to 75mm
  • The quintuple reversed nosepiece revolves with click stop
  • Includes: blue and green filter, 1 x spare bulb, 2 x spare fuses, immersion oil 10mL, soft rubber eyecups, dust cover and instruction manual
  • Images may illustrate the microscope with Coolpix digital camera accessory: please note this is an optional extra and is NOT supplied as standard. For details, please contact Customer Service.
Specifications

Specifications

Filter
32 mm
1 Ea.
Ground
FB69075 - FB69088
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers