missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FGFR3, His Tag Recombinant Protein

FGFR3 is a member of the FGFR family of receptor tyrosine kinases.

Brand:  Invitrogen™ PR4631B

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15431204

  • 13300.00 DKK / 100µg
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The FGFR family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy.

Specifications

Specifications

NP_000133
49.8 kDa
FGFR3, CEK2, JTK4
Human
His Tag
Tyrosine Kinases
Baculovirus
Recombinant, Catalytic Domain, Cytoplasmic
Kinases & Inhibitors
Kinase Assay
100μg
FGF Receptor Kinase Family
Recombinant
Storage Buffer: 50mM Tris, pH 7.5, 100mM NaCl, 0.2mM EDTA, 0.05% Triton X-100, 5mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Dry Ice
Kinases & Inhibitors
Liquid
Oncology, Signal Transduction
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers