missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FGFR3, His Tag Recombinant Protein
FGFR3 is a member of the FGFR family of receptor tyrosine kinases.
4385.00 DKK - 57500.00 DKK
Specifications
Accession Number | NP_000133 |
---|---|
Gene Alias | FGFR3, CEK2, JTK4 |
KinaseFamily | FGF Receptor Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15431204
|
Invitrogen™
PR4631B |
100μg |
13300.00 DKK
100µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10352163
|
Invitrogen™
PV4107 |
1mg |
57500.00 DKK
1mg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11569326
|
Invitrogen™
PV3145 |
10 μg |
4385.00 DKK
10µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The FGFR family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy.
Specifications
NP_000133 | |
FGF Receptor Kinase Family | |
Recombinant | |
Baculovirus |
FGFR3, CEK2, JTK4 | |
Human | |
His Tag |